missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human CP110 (aa 291-404) Control Fragment Recombinant Protein

Product Code. 30180464
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30180464

Brand: Invitrogen™ RP98222

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (75%), Rat (75%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-58775 (PA5-58775. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Necessary for centrosome duplication at different stages of procentriole formation. Acts as a key negative regulator of ciliogenesis in collaboration with CEP97 by capping the mother centriole thereby preventing cilia formation (PubMed:17719545 PubMed:17681131, PubMed:23486064). Also involved in promoting ciliogenesis. May play a role in the assembly of the mother centriole subdistal appendages (SDA) thereby effecting the fusion of recycling endosomes to basal bodies during cilia formation. Required for correct spindle formation and has a role in regulating cytokinesis and genome stability via cooperation with CALM1 and CETN2 (PubMed:16760425). [UniProt]
TRUSTED_SUSTAINABILITY

Specifications

Accession Number O43303
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 9738
Name Human CP110 (aa 291-404) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 6330503K22Rik; AA415922; AI427129; AW557948; CCP110; centriolar coiled coil protein 110; centriolar coiled coil protein 110 kDa; centriolar coiled-coil protein 110; centriolar coiled-coil protein 110 kDa; centriolar coiled-coil protein of 110 kDa; Centriolin; centrosomal protein CP110; centrosomal protein of 110 kDa; CEP1; Cep110; CNTRL; CP110; FAN; Kiaa0419; RGD1310509
Common Name CP110
Gene Symbol CCP110
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence DKPSLNKSNVLLQGASTQASSMSMPVLASFSKVDIPIRTGHPTVLESNSDFKVIPTFVTENNVIKSLTGSYAKLPSPEPSMSPKMHRRRSRTSSACHILINNPINACELSPKGK
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.