missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human CP native Protein from human plasma
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
Description
The protein encoded by this gene is a metalloprotein that binds most of the copper in plasma and is involved in the peroxidation of Fe(II)transferrin to Fe(III) transferrin. Mutations in this gene cause aceruloplasminemia, which results in iron accumulation and tissue damage, and is associated with diabetes and neurologic abnormalities. [provided by RefSeq]
Specifications
Specifications
| For Use With (Application) | SDS-PAGE |
| Formulation | Lyophilized from 50mM potassium phosphate, 100mM KCl, pH 6.8 (20mM E-amino caproic acid, 5mM EDTA) |
| Gene ID (Entrez) | 1356 |
| Molecular Weight (g/mol) | 132kDa |
| Name | CP (Human) Native Protein |
| Preparation Method | Native protein purified from human plasma |
| Quantity | 1 mg |
| Storage Requirements | Store at -20°C on dry atmosphere. Aliquot to avoid repeated freezing and thawing. |
| Regulatory Status | RUO |
| Gene Alias | CP-2 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction