missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human COX7A2L (aa 3-76) Control Fragment Recombinant Protein

Codice prodotto. 30204820
Click to view available options
Quantity:
100 μL
Dimensione della confezione:
100µL
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Codice prodotto. 30204820

Marca: Invitrogen™ RP106919

Please to purchase this item. Need a web account? Register with us today!

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (85%), Rat (85%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-66237 (PA5-66237. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

The cytochrome c oxidase (COX) family of proteins function as the final electron donor in the respiratory chain to drive a proton gradient across the inner mitochondrial membrane, ultimately resulting in the production of water. The mammalian COX apoenzyme is a dimer, with each monomer consisting of 13 subunits, some of which are mitochondrial and some of which are nuclear. COX7a2 (cytochrome c oxidase subunit VIIa polypeptide 2), also known as COX7AL or COX7AL1, is an 83 amino acid protein that localizes to the inner mitochondrial membrane and exists as a component of the COX complex, playing an important role in electron transport. COX7a2L (cytochrome c oxidase subunit 7A-related protein), also known as COX7AR or COX7RP, is an inner mitochondrial membrane protein that consists of 114 amino acids and is induced by estrogen.
TRUSTED_SUSTAINABILITY

Specifica

Accession Number O14548
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 9167
Name Human COX7A2L (aa 3-76) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias CO x 7A2L; CO x 7 AR; CO x 7 A-related protein; CO x 7 RP; cytochrome c oxidase subunit 7A2 like; cytochrome c oxidase subunit 7 A-related protein, mitochondrial; cytochrome c oxidase subunit VIIa polypeptide 2 like; cytochrome c oxidase subunit VIIa polypeptide 2-like; cytochrome c oxidase subunit VIIa-related protein; cytochrome c oxidase subunit VII-related protein; EB1; estrogen receptor binding CpG island; Scaf1; SIG81; SIG-81; Silg81; Silica-induced gene 81 protein; supercomplex assembly factor I
Common Name COX7A2L
Gene Symbol Cox7a2l
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence YKFSGFTQKLAGAWASEAYSPQGLKPVVSTEAPPIIFATPTKLTSDSTVYDYAGKNKVPELQKFFQKADGVPVY
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Vedi altri risultati Mostra meno risultati
Correzione del contenuto del prodotto

Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.

Titolo del prodotto

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.

Grazie per averci aiutato a migliorare il nostro sito web. Il vostro feedback è stato inviato