missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human COX6C Control Fragment Recombinant Protein

Product Code. 30210064
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30210064

Brand: Invitrogen™ RP102541

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (82%), Rat (82%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-53141 (PA5-53141. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Cytochrome c oxidase, the terminal enzyme of the mitochondrial respiratory chain, catalyzes the electron transfer from reduced cytochrome c to oxygen. It is a heteromeric complex consisting of 3 catalytic subunits encoded by mitochondrial genes and multiple structural subunits encoded by nuclear genes. The mitochondrially-encoded subunits function in electron transfer, and the nuclear-encoded subunits may be involved in the regulation and assembly of the complex. This nuclear gene encodes subunit VIc, which has 77% amino acid sequence identity with mouse subunit VIc. This gene is up-regulated in prostate cancer cells. A pseudogene has been found on chromosomes 16p12.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P09669
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 1345
Name Human COX6C Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias COVIc; Co x 6 c; Co x 6c2; COX-VIc; cytochrome c oxidase polypeptide VIc; Cytochrome c oxidase polypeptide VIc-2; cytochrome c oxidase subunit 6 C; Cytochrome c oxidase subunit 6 C-2; cytochrome c oxidase subunit VIc; cytochrome c oxidase subunit VIc preprotein; cytochrome c oxidase, subunit VIc; cytochrome oxidase subunit VIc; EC 1.9.3.1
Common Name COX6C
Gene Symbol COX6C
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence EPRKKAYADFYRNYDVMKDFEDMRKTGT
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.