missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human COX-1 Antibody, R&D Systems™

Rat Monoclonal Antibody
£146.00 - £346.00
Specifications
| Antigen | COX-1 |
|---|---|
| Clone | 345305 |
| Concentration | LYOPH |
| Dilution | Western Blot 1 μg/mL, Flow Cytometry 0.25 μg/10^6 cells, CyTOF-ready |
| Applications | Western Blot, Flow Cytometry, CyTOF |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18710928
![]() |
R&D Systems
MAB3740-SP |
25 μg |
£146.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18182711
![]() |
R&D Systems
MAB3740 |
100 μg |
£346.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
COX-1 Monoclonal specifically detects COX-1 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.Specifications
| COX-1 | |
| LYOPH | |
| Western Blot, Flow Cytometry, CyTOF | |
| Unconjugated | |
| Rat | |
| Human | |
| P23219 | |
| 5742 | |
| E. coli-derived recombinant human COX-1 Ala24-Leu599 Accession # P23219 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human COX-1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human Semaphorin 3A, 3B, 6B, 6C, 6D, recombinant mouse Semaphorin 3A, 3B, 3E, 3F, 6A, or 7A is observed. |
| 345305 | |
| Western Blot 1 μg/mL, Flow Cytometry 0.25 μg/10^6 cells, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| COX1, COX-1, COX1PGG/HS, COX3, Cyclooxygenase-1, EC 1.14.99.1, PCOX1, PGH synthase 1, PGHS1, PGHS-1PHS1, PHS 1, prostaglandin G/H synthase 1, prostaglandin G/H synthase and cyclooxygenase, Prostaglandin H2 synthase 1, Prostaglandin-endoperoxide synthase 1, prostaglandin-endoperoxide synthase 1 (prostaglandin G/H synthase andcyclooxygenase), PTGHS, PTGS1 | |
| PTGS1 | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title