missing translation for 'onlineSavingsMsg'
Learn More

Human Coronavirus Spike S1 Rabbit anti-HCoV-229E, Polyclonal, Novus Biologicals™

Product Code. 18369314 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
0.5 mg
Unit Size:
0.1mg
0.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18369314 0.1 mg 0.1mg
18356914 0.5 mg 0.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18369314 Supplier Novus Biologicals Supplier No. NBP3148620.1MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Human Coronavirus Spike S1 Polyclonal antibody specifically detects Human Coronavirus Spike S1 in HCoV-229E samples. It is validated for ELISA, Immunomicroscopy
TRUSTED_SUSTAINABILITY

Specifications

Antigen Human Coronavirus Spike S1
Applications ELISA, Immunomicroscopy
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution ELISA, Electron Microscopy
Formulation PBS, 0.2 um filtered.
Host Species Rabbit
Immunogen Recombinant HCoV-229E Spike (S1)
Purification Method Antigen Affinity-purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species HCoV-229E
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.