missing translation for 'onlineSavingsMsg'
Learn More

Human Coronavirus Spike Protein Antibody (610), Alexa Fluor™ 700, Novus Biologicals™

Product Code. 18341777 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18341777 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18341777 Supplier Novus Biologicals Supplier No. NBP314663AF700

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Human Coronavirus Spike Protein Monoclonal antibody specifically detects Human Coronavirus Spike Protein in HCoV-NL63 samples. It is validated for ELISA, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen Human Coronavirus Spike Protein
Applications ELISA, Immunofluorescence
Classification Monoclonal
Clone 610
Conjugate Alexa Fluor 700
Dilution ELISA, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias HKU1, Spike glycoprotein
Host Species Rabbit
Immunogen This product is a recombinant monoclonal antibody expressed from HEK293 cells.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3200426
Target Species HCoV-NL63
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.