missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Coronavirus Spike Protein Antibody (07), DyLight 488, Novus Biologicals™
Shop All Bio Techne ProductsDescription
Human Coronavirus Spike Protein Monoclonal antibody specifically detects Human Coronavirus Spike Protein in HCoV-HKU1 samples. It is validated for ELISA, Immunofluorescence
Specifications
Specifications
| Antigen | Human Coronavirus Spike Protein |
| Applications | ELISA, Immunofluorescence |
| Classification | Monoclonal |
| Clone | 7 |
| Conjugate | DyLight 488 |
| Dilution | ELISA, Immunocytochemistry/Immunofluorescence |
| Formulation | 50mM Sodium Borate |
| Gene Alias | HKU1, Spike glycoprotein |
| Host Species | Mouse |
| Immunogen | This product is a recombinant monoclonal antibody expressed from HEK293 cells. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?