missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Coronavirus Nucleocapsid Protein Rabbit anti-HCoV-OC43, Polyclonal, Novus Biologicals™
Shop All Bio Techne ProductsDescription
Human Coronavirus Nucleocapsid Protein Polyclonal antibody specifically detects Human Coronavirus Nucleocapsid Protein in HCoV-OC43 samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | Human Coronavirus Nucleocapsid Protein |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS, 50% glycerol, pH7.3 |
| Host Species | Rabbit |
| Immunogen | A synthetic peptide corresponding to a sequence within amino acids 100-200 of coronavirus Nucleoprotein (YP_009555245.1). |
| Purification Method | Affinity purified |
| Quantity | 20 μg |
| Regulatory Status | RUO |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?