missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Cornulin Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF3607-SP
This item is not returnable.
View return policy
Description
Cornulin Polyclonal specifically detects Cornulin in Human samples. It is validated for Western Blot.
Specifications
| Cornulin | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| 53 kDa squamous epithelial-induced stress protein, 58 kDa heat shock protein, C1orf10, chromosome 1 open reading frame 10, cornulin, CRNN, DRC1, Gm1015, PDRC1, PDRC1,53 kDa putative calcium-binding protein, SEP53, SEP53C1orf10, Squamous epithelial heat shock protein 53, Tumor-related protein | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 49860 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| Q9UBG3 | |
| CRNN | |
| E. coli-derived recombinant human Cornulin Met1-Ser140 Accession # Q9UBG3 | |
| 25 μg | |
| Primary | |
| Detects human Cornulin in direct ELISAs and Western blots. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction