missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Corin Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB2209-100
This item is not returnable.
View return policy
Description
Corin Monoclonal specifically detects Corin in Human samples. It is validated for ELISA, Immunocytochemistry.
Specifications
| Corin | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| ATC2, atrial natriuteric peptide-converting enzyme, Corin, corin, serine peptidase, EC 3.4.21, EC 3.4.21.-, heart specific serine proteinase, Heart-specific serine proteinase ATC2, MGC119742, pro-ANP-convertase, Pro-ANP-converting enzyme, PRSC, serine protease, Transmembrane protease serine 10 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 10699 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
| Western Blot, Immunocytochemistry | |
| 231443 | |
| Western Blot 1 ug/mL, Immunocytochemistry 8-25 ug/mL | |
| Q9Y5Q5 | |
| CORIN | |
| Mouse myeloma cell line NS0-derived recombinant human Corin Ser67-Asn1042 Accession # Q9Y5Q5 | |
| 100 μg | |
| Primary | |
| Detects human Corin in direct ELISAs and Western blots. | |
| Human | |
| Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction