missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human COPS4 (aa 225-316) Control Fragment Recombinant Protein

Product Code. 30196741
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30196741

Brand: Invitrogen™ RP96659

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (100%), Rat (100%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-57863 (PA5-57863. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

This gene encodes one of eight subunits composing COP9 signalosome, a highly conserved protein complex that functions as an important regulator in multiple signaling pathways. The structure and function of COP9 signalosome is similar to that of the 19S regulatory particle of 26S proteasome. COP9 signalosome has been shown to interact with SCF-type E3 ubiquitin ligases and act as a positive regulator of E3 ubiquitin ligases. Two transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9BT78
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 51138
Name Human COPS4 (aa 225-316) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias AW208976; COP9 (constitutive photomorphogenic) homolog, subunit 4; COP9 (constitutive photomorphogenic) homolog, subunit 4 (Arabidopsis thaliana); COP9 (constitutive photomorphogenic), subunit 4; COP9 complex S4; COP9 constitutive photomorphogenic homolog subunit 4; COP9 constitutive photomorphogenic-like protein subunit 4; COP9 signalosome complex subunit 4; COP9 signalosome subunit 4; COPS4; CSN4; D5Ertd774e; JAB1-containing signalosome subunit 4; SGN4; signalosome subunit 4; testis tissue sperm-binding protein Li 42 A
Common Name COPS4
Gene Symbol COPS4
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence ALKHALHCTILASAGQQRSRMLATLFKDERCQQLAAYGILEKMYLDRIIRGNQLQEFAAMLMPHQKATTADGSSILDRAVIEHNLLSASKLY
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.