missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human COP1 (aa 173-197) Control Fragment Recombinant Protein
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Description
Highest antigen sequence indentity to the following orthologs: Mouse (48%), Rat (48%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-62890 (PA5-62890. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
Caspase inhibitor. Acts as a regulator of procaspase-1/CASP1 activation implicated in the regulation of the proteolytic maturation of pro-interleukin-1 beta (IL1B) and its release during inflammation. Inhibits the release of IL1B in response to LPS in monocytes. Also induces NF-kappa-B activation during the pro-inflammatory cytokine response. Also able to inhibit CASP1-mediated neuronal cell death, TNF-alpha, hypoxia-, UV-, and staurosporine-mediated cell death but not ER stress-mediated cell death. Acts by preventing activation of caspases CASP1 and CASP4, possibly by preventing the interaction between CASP1 and RIPK2. [UniProt]
Specifications
Specifications
| Accession Number | Q5EG05 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 114769 |
| Name | Human COP1 (aa 173-197) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | CARD only domain-containing protein 1; CARD only protein; CARD16; CARD-only protein 1; caspase recruitment domain family member 16; caspase recruitment domain family, member 16; caspase recruitment domain-containing protein 16; Caspase recruitment domain-only protein 1; caspase-1 dominant-negative inhibitor pseudo-ICE; caspase-1 inhibitor COP; Constitutive photomorphogenesis protein 1 homolog; constitutive photomorphogenic protein 1; COP; COP1; E3 ubiquitin-protein ligase COP1; E3 ubiquitin-protein ligase RFWD2; hCOP1; Pseudo interleukin-1 beta converting enzyme; pseudo interleukin-1 beta converting enzyme; PSEUDO-ICE; pseudo-IL1B-converting enzyme; putative ubiquitin ligase COP1; RFWD2; ring finger and WD repeat domain 2; ring finger and WD repeat domain 2, E3 ubiquitin protein ligase; RING finger and WD repeat domain protein 2; RING finger protein 200; RING-type E3 ubiquitin transferase RFWD2; RNF200 |
| Common Name | COP1 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction