Learn More
R&D Systems™ Human Connexin 32/GJB1 (NP_000157) VersaClone cDNA
Product Code. 18732548
Change view
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
Product Code. Quantity unitSize
18732548 10 μg 10µg
1 options
Product Code. 18732548 Supplier R&D Systems™ Supplier No. RDC1026
Gap junction protein, beta 1, 32kDa
The Human Connexin 32/GJB1 (NP_000157) VersaClone cDNA clone contains the complete ORF for Connexin 32/GJB1, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The Connexin 32/GJB1 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
Order Info
Ships at ambient temperature.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Quantity 10 μg
For Reasearch Use Only; Not for Use in Humans