missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Complement Factor MASP3 Catalytic Domain Biotinylated Antibody, R&D Systems™
Goat Polyclonal Antibody
Brand: R&D Systems BAF1724
This item is not returnable.
View return policy
Description
Complement Factor MASP3/MASP1 Polyclonal specifically detects Complement Factor MASP3/MASP1 in Human samples. It is validated for Western Blot.
Specifications
| Complement Factor MASP3/MASP1 | |
| Polyclonal | |
| Biotin | |
| NP_624302 | |
| MASP1 | |
| Mouse myeloma cell line NS0-derived recombinant human Complement Factor MASP3 Catalytic Domain Ile450-Val721 Accession # NP_624302 | |
| 50 μg | |
| Primary | |
| Detects human Complement Factor MASP3 Catalytic Domain in Western blots. | |
| Human | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| Complement factor MASP-3, Complement-activating component of Ra-reactive factor, CRARF1, CRARFMAP1, DKFZp686I01199, EC 3.4.21, EC 3.4.21.-, EC 3.4.21.104, EC 3.4.21.42, FLJ26383, mannan-binding lectin serine peptidase 1 (C4/C2 activating component ofRa-reactive factor), mannan-binding lectin serine protease 1, mannan-binding lectin serine protease 1 (C4/C2 activating component ofRa-reactive factor), Mannose-binding lectin-associated serine protease 1, Mannose-binding protein-associated serine protease, MAp44, MASP, MASP1, MASP-1, MASP3, MGC126283, MGC126284, PRSS5, Ra-reactive factor serine protease p100, RaRF, Serine protease 5 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 5648 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction