missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human Complement Factor H DuoSet ELISA
Description
The Human Complement Factor H DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies human Complement Factor H.
Specifications
Specifications
| Quantity | 15 x 96-well Plates |
| Conjugate | Biotin |
| Assay Range | 0.21 - 150 ng/mL |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 3075 |
| Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?