missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Complement Component C9 Antibody, R&D Systems™
Sheep Polyclonal Antibody
Brand: R&D Systems AF8126
This item is not returnable.
View return policy
Description
Complement Component C9 Polyclonal specifically detects Complement Component C9 in Human samples. It is validated for Western Blot.
Specifications
| Complement Component C9 | |
| Polyclonal | |
| Unconjugated | |
| P02748 | |
| C9 | |
| Human embryonic kidney cell line HEK293-derived human Complement Component C9 Gln22-Lys559 (predicted) Accession # P02748 | |
| 100 μg | |
| Primary | |
| Detects human Complement Component C9 in directs ELISAs and Western blots. | |
| Human | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 0.2 ug/mL | |
| complement component 9, complement component C9 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 735 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur