missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Complement Component C3aR Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB5995-SP
This item is not returnable.
View return policy
Description
Complement Component C3aR Monoclonal specifically detects Complement Component C3aR in Human samples. It is validated for Flow Cytometry, CyTOF-ready.
Specifications
| Complement Component C3aR | |
| Monoclonal | |
| LYOPH | |
| Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
| Q16581 | |
| C3AR1 | |
| NS0 mouse myeloma cell line transfected with human C3a R Met1-Val482 Accession # Q16581 | |
| 25 μg | |
| Primary | |
| Detects human Complement Component C3a R in direct ELISAs. | |
| Human | |
| Purified |
| Flow Cytometry, CyTOF | |
| 534625 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| AZ3B, AZ3BC3a anaphylatoxin chemotactic receptor, C3a-R, C3AR, C3AR1, C3ARcomplement component 3 receptor 1, C3R1, complement component 3a receptor 1, Complement Component C3a R, Complement Component C3aR, HNFAG09 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 719 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction