missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Complement Component C3aR Alexa Fluor 488-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB5995G-100UG
This item is not returnable.
View return policy
Description
Complement Component C3aR Monoclonal specifically detects Complement Component C3aR in Human samples. It is validated for Flow Cytometry.
Specifications
| Complement Component C3aR | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| Q16581 | |
| C3AR1 | |
| NS0 mouse myeloma cell line transfected with human C3a R Met1-Val482 Accession # Q16581 | |
| RUO | |
| 719 | |
| Human | |
| IgG2a |
| Flow Cytometry | |
| 534625 | |
| Alexa Fluor 488 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| AZ3B, AZ3BC3a anaphylatoxin chemotactic receptor, C3a-R, C3AR, C3AR1, C3ARcomplement component 3 receptor 1, C3R1, complement component 3a receptor 1, Complement Component C3a R, Complement Component C3aR, HNFAG09 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human Complement Component C3a R in direct ELISAs. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction