missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Complement Component C1s (Research Grade Sutimlimab Biosimilar) Antibody, Novus Biologicals™

Human Monoclonal Antibody
£171.00 - £447.00
Specifications
| Antigen | Complement Component C1s |
|---|---|
| Clone | Hu222 |
| Dilution | Flow Cytometry 0.25 μg/10^6 cells, ELISA |
| Applications | ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30130443
|
R&D Systems
MAB11489-100 |
100 μg |
£447.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30130420
![]() |
R&D Systems
MAB11489-SP |
25 μg |
£171.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Complement Component C1s Monoclonal antibody specifically detects Complement Component C1s in Human samples. It is validated for ELISASpecifications
| Complement Component C1s | |
| Flow Cytometry 0.25 μg/10^6 cells, ELISA | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| basic proline-rich peptide IB-1, C1 esterase, C1S, complement C1s subcomponent, Complement component 1 subcomponent s, complement component 1, s subcomponent, EC 3.4.21, EC 3.4.21.42, FLJ44757 | |
| NS0 mouse myeloma cell line transfected with human CD38, Met1-Ile300, Accession # P28907 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
| Hu222 | |
| ELISA | |
| Unconjugated | |
| Human | |
| Human | |
| P28907 | |
| 716 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title