missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Complement Component C1r Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 2 publications
Brand: R&D Systems AF1807
Additional Details : Weight : 0.00970kg
Description
Complement Component C1r Polyclonal specifically detects Complement Component C1r in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
Complement Component C1r | |
Polyclonal | |
Unconjugated | |
NP_001724 | |
C1R | |
Mouse myeloma cell line NS0-derived recombinant human Complement Component C1r Ser18-Asp705 Accession # NP_001724 | |
100 μg | |
Primary | |
Detects human Complement Component C1r in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human Complement Component C1s is observed. | |
Human | |
IgG |
Western Blot, Immunoprecipitation | |
LYOPH | |
Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL | |
C1r, complement C1r subcomponent, Complement component 1 subcomponent r, complement component 1, r subcomponent, EC 3.4.21, EC 3.4.21.41 | |
Goat | |
Affinity Purified | |
RUO | |
715 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only