missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human Common gamma Chain/IL-2 R gamma DuoSet ELISA

Product Code. 18746516
Click to view available options
Quantity:
1 Kit (for 5 Plates)
Unit Size:
Pack of 5
This item is not returnable. View return policy

Product Code. 18746516

Brand: R&D Systems™ DY28405

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

Capture antibody, detection antibody, recombinant standard, streptavidin conjugated to horseradish-peroxidase (Streptavidin-HRP) and development protocols

For the determination of natural and recombinant common gamma chain in humans. R&D Systems Human Common gamma Chain/IL-2 R gamma DuoSet ELISA Kit is a solid phase sandwich ELISA suitable for the analysis of cell culture supernate, serum, and plasma samples.

The Human Common gamma Chain/IL-2 R gamma DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies human Common gamma Chain/IL-2 R gamma.
TRUSTED_SUSTAINABILITY

Specifications

Description Common gamma Chain/IL-2 R gamma
Product Type ELISA Kit
Quantity 1 Kit (for 5 Plates)
Species Human
Cross Reactivity Not observed
Format 96-well Strip Plate
For Use With (Application) ELISA
Sample Type Cell Culture Supernatant, Serum, Plasma
Kit Contents Capture antibody, Detection antibody, Recombinant standard, Streptavidin conjugated to horseradish-peroxidase (Streptavidin-HRP) and development protocols
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.