missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Common beta Chain Antibody, R&D Systems™

Mouse Monoclonal Antibody
£171.00 - £496.00
Specifications
| Antigen | Common beta Chain |
|---|---|
| Clone | 140516 |
| Dilution | Western Blot 1 ug/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18715759
![]() |
R&D Systems
MAB906-SP |
25 μg |
£171.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18113391
|
R&D Systems
MAB906 |
500 μg |
£496.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Common beta Chain Monoclonal specifically detects Common beta Chain in Human samples. It is validated for Western Blot.Specifications
| Common beta Chain | |
| Western Blot 1 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| beta, low-affinity, CD131, CD131 antigen, CDw131, colony stimulating factor 2 receptor, beta, low-affinity(granulocyte-macrophage), CSF2RB, cytokine receptor common subunit beta, GM-CSF R beta, GM-CSF/IL-3/IL-5 receptor common beta subunit, GM-CSF/IL-3/IL-5 receptor common beta-chain, IL-3 R beta, IL3RB, IL-5 R beta, interleukin 3 receptor/granulocyte-macrophage colony stimulating factor 3receptor, beta (high affinity) | |
| CSF2RB | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 140516 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| P32927 | |
| 1439 | |
| Mouse myeloma cell line NS0-derived recombinant human Common beta Chain Gly23-Trp443 Accession # P32927 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human Common beta Chain in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) G-CSF R or rhM-CSF R is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title