missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
The Human Collagen VI ELISA Kit (Chemiluminescence) measures human Collagen VI in sandwich elisa.
Specifications
Specifications
| Assay Range | 1.56-100 ng/mL |
| Assay Sensitivity | 0.94 ng/mL |
| Cross Reactivity | This kit recognizes Human COL6 in samples. No significant cross-reactivity or interference between Human COL6 and analogues was observed. |
| Product Type | ELISA Kit (Chemiluminescence) |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 1291 |
| Gene Symbol | COL6A1 |
| Host Species | Human |
| Quantity | 1 Kit |
| Storage Requirements | Storage of components varies. See protocol for specific instructions. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?