missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
The Human COL15A1 ELISA Kit (Chemiluminescence) measures human COL15A1 in serum, plasma and other biological fluids.
Specifications
Specifications
| Assay Range | 62.50-4000 pg/mL |
| Assay Sensitivity | 37.50 pg/mL |
| Cross Reactivity | This kit recognizes Human COL15 in samples. No significant cross-reactivity or interference between Human COL15 and analogues was observed. |
| Product Type | ELISA Kit (Chemiluminescence) |
| Sample Type | Biological Fluids, Plasma, Serum |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 1306 |
| Gene Symbol | COL15A1 |
| Host Species | Human |
| Quantity | 1 Kit |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?