missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human COCO Antibody, R&D Systems™

Mouse Monoclonal Antibody
Marca: R&D Systems MAB30471-SP
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
COCO/DAND5 Monoclonal specifically detects COCO/DAND5 in Human samples. It is validated for ELISA Capture (Matched Antibody Pair).
Especificaciones
| COCO/DAND5 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CER2CRL2, Cerberus 2, Cerberus like-2, cerberus-like 2, Cerberus-like protein 2, cerl-2, CERL2, CKTSF1B3, CKTSF1B3MGC126849, COCO, Cysteine knot superfamily 1, BMP antagonist 3, DAN domain family member 5, DAN domain family, member 5, DAND5, Dante, DTE, FLJ38607, GREM3, GREM3cerberus 2, Gremlin-3, SP1 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 199699 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| ELISA | |
| 322209 | |
| ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| Q8N907 | |
| DAND5 | |
| E. coli-derived recombinant human COCO Arg23-Ala189 Accession # Q8N907 | |
| 25 μg | |
| Primary | |
| Detects human COCO in ELISAs. In sandwich immunoassays, no cross-reactivity or interference with recombinant human (rh) Activin A, rhBMP-4, rhDAN, or recombinant mouse COCO is observed. | |
| Human | |
| Purified |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido