missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human Coagulation Factor XIV/Protein C ELISA Kit

Product Code. 17174393
Change view
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
Unit Size:
Each
Pack of 96
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17174393 96 Tests Pack of 96
17114869 5 x 96 Tests Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17174393 Supplier Invitrogen™ Supplier No. EH119RB

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ELISA

Human Coagulation Factor XIV/Protein C quantitates human Coagulation Factor XIV/Protein C in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human Coagulation Factor XIV/Protein C.

Protein C is a vitamin K-dependent plasma glycoprotein that is cleaved to its activated form by the thrombin-thrombomodulin complex. Protein C in its activated form contains a serine protease domain and functions in degradation of the activated forms of coagulation factors V and VIII. Protein C is a vitamin K-dependent serine protease produced in the liver and made up of 2 polypeptide chains. The 62 kDa proenzyme is activated by thrombin bound to an endothelial surface receptor and the active enzyme cleaves factor Va and VIIIa and thus inhibits blood coagulation. The molecular weight of the active enzyme is 55 kDa and the normal concentrations in human plasma is approximately 1-3 ng/mL because of the very fast turnover, the proenzyme concentration is approximately 3 μg/mL. Mutations in the Protein C gene have been associated with thrombophilia due to protein C deficiency, neonatal purpura fulminans, and recurrent venous thrombosis.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Activation peptide; anticoagulant protein C; APC; Autoprothrombin IIA; blood coagulation factor XIV; EDQVDPRLIDGK; inactivator of coagulation factors Va, VIII; PC; prepro-protein C; Proc; PROC1; protein C; protein C (inactivator of coagulation factors Va and VIIIa); protein C prepropeptide; protein C, inactivator of coagulation factors Va and VIIIa; THPH3; THPH4; type I protein C; vitamin K-dependent protein C; Vitamin K-dependent protein C heavy chain; Vitamin K-dependent protein C light chain
Assay Range 0.82 to 20 ng/mL
Assay Sensitivity 0.82 ng/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 5624
Gene Symbol PROC
Interassay CV <12%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Storage Requirements 2°C to 8°C
Target Protein C
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 45 mins
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.