missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Coagulation Factor X Antibody, R&D Systems™

Mouse Monoclonal Antibody
£167.00 - £523.00
Specifications
| Antigen | Coagulation Factor X |
|---|---|
| Clone | 156106 |
| Concentration | LYOPH |
| Dilution | Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL |
| Applications | Western Blot, Immunoprecipitation |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18792827
![]() |
R&D Systems
MAB1063-SP |
25 μg |
£167.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18172611
![]() |
R&D Systems
MAB1063 |
500 μg |
£523.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Coagulation Factor X Monoclonal specifically detects Coagulation Factor X in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
| Coagulation Factor X | |
| LYOPH | |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| Mouse | |
| Human | |
| P00742 | |
| 2159 | |
| Mouse myeloma cell line NS0-derived recombinant human Coagulation Factor X Leu24-Lys488 (predicted) Accession # P00742 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human Coagulation Factor X in direct ELISAs and Western blots. It recognizes both Factor X and Factor Xa. |
| 156106 | |
| Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Cf10, coagulation factor X, EC 3.4.21, EC 3.4.21.6, F10, factor Xa, FX, FXa, prothrombinase, Stuart Factor, Stuart-Prower factor | |
| F10 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title