missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human Coagulation Factor VII ELISA Kit
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
Unit Size:
Each
Pack of 96
Description
Human Coagulation Factor VII quantitates human Coagulation Factor VII in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human Coagulation Factor VII.
Coagulation factor VII is a vitamin K-dependent factor essential for hemostasis. This factor circulates in the blood in a zymogen form, and is converted to an active form by either factor IXa, factor Xa, factor XIIa, or thrombin by minor proteolysis. Upon activation of the factor VII, a heavy chain containing a catalytic domain and a light chain containing 2 EGF-like domains are generated, and two chains are held together by a disulfide bond. In the presence of factor III and calcium ions, the activated factor then further activates the coagulation cascade by converting factor IX to factor IXa and/or factor X to factor Xa. Defects in F7 production can cause coagulopathy.
Specifications
Specifications
| Accession Number | AI132620; Cf7; Coagulation factor VII; coagulation factor VII (serum prothrombin conversion accelerator); eptacog alfa; F7; Factor 7; Factor VII heavy chain; Factor VII light chain; Factor7; FVII; FVII coagulation protein; proconvertin; Serum prothrombin conversion accelerator; SPCA |
| Assay Range | 0.4 to 100 ng/mL |
| Assay Sensitivity | 0.4 ng/mL |
| Conjugate | HRP |
| Product Type | ELISA |
| Sample Type | Plasma, Serum, Supernatant |
| For Use With (Equipment) | Colorimetric Microplate Reader |
| Gene ID (Entrez) | 2155 |
| Gene Symbol | F7 |
| Interassay CV | <12% |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction