missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Coagulation Factor V Antibody, R&D Systems™

Mouse Monoclonal Antibody
£164.00 - £397.00
Specifications
| Antigen | Factor V |
|---|---|
| Clone | 986237 |
| Dilution | ELISA |
| Applications | ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18783157
![]() |
R&D Systems
MAB98562-SP |
25 μg |
£164.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18701447
![]() |
R&D Systems
MAB98562-100 |
100 μg |
£397.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Factor V Monoclonal specifically detects Factor V in Human samples. It is validated for ELISA.Specifications
| Factor V | |
| ELISA | |
| Monoclonal | |
| Supernatant | |
| Human | |
| Activated protein C cofactor, Cf5, Cf-5, Coagulation Factor 5, coagulation factor V, coagulation factor V (proaccelerin, labile factor), coagulation factor V jinjiang A2 domain, F5, Factor V, factor V Leiden, FVL, PCCF, Proaccelerin, labile factor, RPRGL1, THPH2 | |
| Human Coagulation Factor V synthetic peptide | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human Coagulation Factor V in direct ELISAs. |
| 986237 | |
| ELISA | |
| Unconjugated | |
| Mouse | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| 2153 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title