missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human Coagulation Factor III/Tissue Factor Quantikine ELISA
Description
The Human Coagulation Factor III/Tissue Factor Quantikine ELISA is a Solid Phase Sandwich ELISA that quantifies human Coagulation Factor III/Tissue Factor in cell culture supernates (50 ul), cell lysates (50 ul), edta plasma (50 ul), heparin plasma (50 ul), citrate plasma (50 ul), urine (50 ul).
Specifications
Specifications
| Assay Range | 7.80 - 500pg/mL |
| Assay Sensitivity | 2.05pg/mL |
| Conjugate | HRP |
| Sample Type | Cell Culture Supernatant, Cell Lysates, Plasma (EDTA, Citrate, Heparin), Urine |
| Specificity | Natural and Recombinant Human Coagulation Factor III |
| Sufficient For | 1 x 96-well Plate |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 2152 |
| Sample Volume | Cell Culture Supernates (50uL),Cell Lysates (50uL),EDTA Plasma (50uL),Heparin Plasma (50uL),Citrate Plasma (50uL),Urine (50uL) |
| Storage Requirements | Store the unopened product at 2° to 8°C. Do not use past expiration date. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?