missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human CNTF, Animal-Free Recombinant Protein, PeproTech®

Product Code. 17813283
Click to view available options
Quantity:
1 mg
100 μg
20 μg
500 μg
Unit Size:
100µg
1mg
20µg
500µg
This item is not returnable. View return policy

Product Code. 17813283

Brand: Gibco™ AF4501320UG

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Recombinant Protein

Recombinant Human CNTF is synthesized as a 199 amino acid polypeptide (22.8 kDa) lacking a hydrophobic N-terminal signal for secretion. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

The protein encoded by this gene is a polypeptide hormone whose actions appear to be restricted to the nervous system where it promotes neurotransmitter synthesis and neurite outgrowth in certain neuronal populations. The protein is a potent survival factor for neurons and oligodendrocytes and may be relevant in reducing tissue destruction during inflammatory attacks. A mutation in this gene, which results in aberrant splicing, leads to ciliary neurotrophic factor deficiency, but this phenotype is not causally related to neurologic disease. A read-through transcript variant composed of ZFP91 and CNTF sequence has been identified, but it is thought to be non-coding. Read-through transcription of ZFP91 and CNTF has also been observed in mouse.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P26441
For Use With (Application) Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 1270
Molecular Weight (g/mol) 22.8 kDa
Name Human CNTF, Animal-Free
Quantity 20 μg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <0.1 EU/ μg
Gene Alias AI429687; Ciliary neurotrophic factor; ciliary neurotropic factor; Cntf; HCNTF; H-CNTF
Common Name CNTF
Gene Symbol CNTF
Biological Activity Determined by its ability to stimulate proliferation of human TF-1 cells using an ED50 concentration range of 50-150 ng/ml
Conjugate Unconjugated
Recombinant Recombinant
Sequence AFTEHSPLTP HRRDLCSRSI WLARKIRSDL TALTQSYVKH QGLNKNINLD SADGMPVAST DQWSQLTQAQ RLQQNLQAYR TFHVLLARLL QDQQVHFTPT QGDFHQAIHT LLLQVAAFAY QIQQLMILLQ YKIPRNQADG MPINVGDGGL FQKKLWGLKV LQQLSQWTVR SIHDLRFISS HQTGIPARGS HYIANNKKM
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.