missing translation for 'onlineSavingsMsg'
Learn More

Human CNP Propeptide Antibody, R&D Systems™
GREENER_CHOICE

Rat Monoclonal Antibody

£146.00 - £346.00

Specifications

Antigen CNP/NPPC
Clone 347625
Concentration LYOPH
Dilution Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL
Applications Western Blot, Immunohistochemistry
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18719128
GREENER_CHOICE
View Documents
R&D Systems
MAB3739-SP
25 μg
£146.00
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18188950
GREENER_CHOICE
View Documents
R&D Systems
MAB3739
100 μg
£346.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

CNP/NPPC Monoclonal specifically detects CNP/NPPC in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications

Specifications

CNP/NPPC
LYOPH
Western Blot, Immunohistochemistry
Unconjugated
Rat
Human
P23582
4880
E. coli-derived recombinant human CNP Lys24-Cys126 Accession # P23582
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Detects human CNP Propeptide in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-react with recombinant human (rh) ANP precursor or rhBNP precursor is observed.
347625
Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL
Monoclonal
Purified
RUO
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
CNP, CNP2, C-type natriuretic peptide, natriuretic peptide C, natriuretic peptide precursor C, NPPC
NPPC
IgG2a
Protein A or G purified from hybridoma culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.