missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human CNOT7 (aa 257-285) Control Fragment Recombinant Protein

Product Code. 30211358
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30211358

Brand: Invitrogen™ RP107696

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (97%), Rat (97%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-67055 (PA5-67055. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

CNOT7 (CCR4-NOT transcription complex subunit 7) has 3'-5' poly(A) exoribonuclease activity for synthetic poly(A) RNA substrate. Its function seems to be partially redundant with that of CNOT8. CNOT7 is a catalytic component of the CCR4-NOT complex, which is one of the major cellular mRNA deadenylases, and is linked to various cellular processes including bulk mRNA degradation, miRNA-mediated repression, translational repression during translational initiation, and general transcription regulation. During miRNA-mediated repression, the complex seems to act as a transcriptional repressor during translational initiation, as well. Additional complex functions may be a consequence of its influence on mRNA expression. CNOT7 associates with members of the BTG family such as TOB1 and BTG2 and is required for their anti-proliferative activity.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9UIV1
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 29883
Name Human CNOT7 (aa 257-285) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias AU022737; BTG1-binding factor 1; CAF1; CAF-1; Caf1a; carbon catabolite repressor protein (CCR4)-associative factor 1; catabolite repressor protein (CCR4)-associative factor 1; CCR4-associated factor 1; CCR4-NOT transcription complex subunit 7; CCR4-NOT transcription complex subunit-like protein; CCR4-NOT transcription complex, subunit 7; CNOT7; fd59c07; fj07d05; hCAF-1; hypothetical protein LOC508055; Pop2; POP2 homolog; wu:fd59c07; wu:fj07d05; zgc:153168
Common Name CNOT7
Gene Symbol CNOT7
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence GHLYGLGSGSSYVQNGTGNAYEEEANKQS
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.