missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human CMYA5 (aa 2280-2356) Control Fragment Recombinant Protein

Product Code. 30200665
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30200665

Brand: Invitrogen™ RP109651

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (40%), Rat (40%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-140296 (PA5-140296. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

SPRYD2, also known as Myospryn, was originally identified as the muscle-specific partner of dysbindin and as a Mef-2 target gene. It is a large scaffolding protein localized to the Z-disc/costamere region of striated muscle. SPRYD2 includes a noncanonical tripartite motif (TRIM-like) that lacks the RING domain but consists of a B-box coiled coil (BBC), fibronectin 3 (FN3) repeats, and SPRY domains. SPRYD2 interacts with desmin and calcineurin, and it has been suggested to play a role in the biogenesis of lysosome and negatively regulates slow-fiber-type transformation and skeletal muscle regeneration. SPRYD2 is dysregulated in Duchenne muscular dystrophy.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q8N3K9
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 202333
Name Human CMYA5 (aa 2280-2356) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 2310076E16Rik; 2310076E21Rik; AA420382; AI504003; C5orf10; cardiomyopathy associated 5; cardiomyopathy-associated protein 5; CMYA5; DTNBP2; Dystrobrevin-binding protein 2; genethonin-3; heart and skeletal muscle-specific and sprouty domain-containing; myospryn; SPRY domain-containing protein 2; SPRYD2; sr553; Srfsd; stretch response protein 553; stretch-response protein 553; stretch-responsive fibronectin protein type 3; Tims; TRIM76; TRIM-like protein; tripartite motif-containing 76; tripartite motif-containing protein 76
Common Name CMYA5
Gene Symbol CMYA5
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence FISEVSREDYGKKEISGDSEEMNINSVVTSADGENLEIQSYSLIGEKLVMEEAKTIVPPHVTDSKRVQKPAIAPPSK
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.