missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human CMPK2 (aa 330-425) Control Fragment Recombinant Protein

Product Code. 30197953
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30197953

Brand: Invitrogen™ RP110205

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (86%), Rat (86%). This recombinant protein control fragment may be used for blocking experiments. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

UMP-CMP kinase 2 (CMPK2) is the first nucleoside monophosphate kinase that has been identified in human mitochondria. It is a component of the salvage pathway for nucleotide synthesis that may participate in terminal differentiation of monocytic cells. There are two distinct domains of CMPK2: the N-terminal domain with unknown function and the C-terminal domain with the kinase function, suggesting that CMPK2 may be a bifunctional protein with other biological functions in addition to its UMP-CMP kinase activity. CMPK2 may participate in dUTP and dCTP synthesis and is responsible for phosphorylation of dCMP and dUMP in mitochondria. CMPK2 is actively involved in macrophage activation and the inflammatory response.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q5EBM0
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 129607
Name Human CMPK2 (aa 330-425) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 1200004E04Rik; Cmpk2; cytidine monophosphate (UMP-CMP) kinase 2, mitochondrial; cytidine/uridine monophosphate kinase 2; cytidylate kinase 2; mitochondrial UMP-CMP kinase; NDK; Nucleoside-diphosphate kinase; TDKI; thymidine monophosphate kinase 2; thymidylate kinase family LPS-inducible member; thymidylate kinase homologue; thymidylate kinase LPS-inducible member; TMPK2; Tyki; UMP/CMP kinase; UMP-CMP kinase 2, mitochondrial; UMP-CMPK2
Common Name CMPK2
Gene Symbol CMPK2
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence DRYWHSTATYAIATEVSGGLQHLPPAHHPVYQWPEDLLKPDLILLLTVSPEERLQRLQGRGMEKTREEAELEANSVFRQKVEMSYQRMENPGCHVV
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.