missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CMG-2/ANTXR2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
£167.00 - £420.00
Specifications
| Antigen | CMG-2/ANTXR2 |
|---|---|
| Clone | 406510 |
| Dilution | ELISA |
| Applications | ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18701778
![]() |
R&D Systems
MAB29401-SP |
25 μg |
£167.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18773529
![]() |
R&D Systems
MAB29401-100 |
100 μg |
£420.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CMG-2/ANTXR2 Monoclonal specifically detects CMG-2/ANTXR2 in Human samples. It is validated for ELISA.Specifications
| CMG-2/ANTXR2 | |
| ELISA | |
| Monoclonal | |
| Supernatant | |
| Human | |
| P58335 | |
| 118429 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 406510 | |
| ELISA | |
| Unconjugated | |
| Mouse | |
| Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2μm filtered solution in PBS. | |
| anthrax toxin receptor 2, ANTXR2, Capillary morphogenesis gene 2 protein, capillary morphogenesis protein 2, cI-35, CMG2, CMG2MGC111533, CMG-2MGC45856, FLJ31074, ISH, JHF, JHS | |
| Mouse myeloma cell line NS0-derived recombinant human CMG-2/ANTXR2 Ser22-Arg200 Accession # P58335 | |
| Primary | |
| Reconstitute at 0.5mg/mL in sterile PBS. | |
| Detects human CMG-2/ANTXR2 in direct ELISAs. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title