missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human CMASP-3 ELISA Kit

Codice prodotto. 17114393
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
Dimensione della confezione:
Each
Pack of 96
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Codice prodotto. 17114393

Marca: Invitrogen™ EH113RB

Please to purchase this item. Need a web account? Register with us today!

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

ELISA

Human CMASP-3 quantitates human CMASP-3 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human CMASP-3.

MBL is a key component in immune response in that it can directly trigger neutralization of invading microorganisms by an Ab-independent mechanism. Mutations of human MBL are associated with immunodeficiency resulting from a reduction in the ability of the mutant MBL to initiate complement fixation. In human, three types of MBL-associated serine proteases, MASP-1, MASP-2 and MASP-3, and a truncated form of MASP-2 (small MBL-associated protein; sMAP or MAp19) complex with MBL to activate the lectin pathway of the complement system. MASP-3 is an alternatively spliced product from the MASP-1 gene. The heavy/A chains are identical between MASP-1 and MASP-3 but the light/B chains are entirely different. Activated MASPs subsequently cleave and activate downstream components of the complement pathway.
TRUSTED_SUSTAINABILITY

Specifica

Accession Number 3 MC1; AW048060; C4/C2 activating component of Ra-reactive factor; CCPII; Complement factor MASP-3; complement-activating component of Ra-reactive factor; Crarf; CRARF1; mannan binding lectin serine peptidase 1; mannan-binding; mannan-binding lectin serine peptidase 1; mannan-binding lectin serine peptidase 1 (C4/C2 activating component of Ra-reactive factor); mannan-binding lectin serine protease 1; Mannan-binding lectin serine protease 1 heavy chain; Mannan-binding lectin serine protease 1 light chain; Mannose-binding lectin-associated serine protease 1; mannose-binding protein associated serine protease-1; mannose-binding protein-associated serine protease; MAP1; MAp44; MASP; Masp1; MASP-1; Masp1/3; MASP3; PRSS5; Ra-reactive factor serine protease p100; RaRF; Serine protease 5; serine protease MASP1
Assay Range 150 to 0.6 ng/mL
Assay Sensitivity 0.6 ng/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 5648
Gene Symbol MASP1
Interassay CV <12%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Storage Requirements 2°C to 8°C
Target MASP1
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 45 mins
Vedi altri risultati Mostra meno risultati
Correzione del contenuto del prodotto

Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.

Titolo del prodotto

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.

Grazie per averci aiutato a migliorare il nostro sito web. Il vostro feedback è stato inviato