missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Clusterin-like 1/CLUL1 Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF3185-SP
This item is not returnable.
View return policy
Description
Clusterin-like 1/CLUL1 Polyclonal specifically detects Clusterin-like 1/CLUL1 in Human samples. It is validated for Western Blot.
Specifications
| Clusterin-like 1/CLUL1 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CLUL1, Clusterin like 1, clusterin-like 1 (retinal), clusterin-like protein 1, RA337M, Retinal-specific clusterin-like protein | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 27098 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| Q15846 | |
| CLUL1 | |
| Mouse myeloma cell line NS0-derived recombinant human Clusterin-like 1/CLUL1 Cys20-Ala442 Accession # Q15846 | |
| 25 μg | |
| Primary | |
| Detects human Clusterin-like 1/CLUL1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 5% cross-reactivity with recombinant human Clusterin is observed. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction