Learn More
Human Clusterin-like 1/CLUL1 Antibody, R&D Systems™
Goat Polyclonal Antibody
Brand:  R&D Systems AF3185
Additional Details : Weight : 0.00970kg
 View more versions of this product
Product Code. 18009041
  • £464.80 / 100µg
Estimated Shipment: 04-06-2024
to see stock.
Description
Description
Clusterin-like 1/CLUL1 Polyclonal specifically detects Clusterin-like 1/CLUL1 in Human samples. It is validated for Western Blot.
Specifications
Specifications
Clusterin-like 1/CLUL1
Polyclonal
Unconjugated
Q15846
CLUL1
Mouse myeloma cell line NS0-derived recombinant human Clusterin-like 1/CLUL1 Cys20-Ala442 Accession # Q15846
100 μg
Primary
Detects human Clusterin-like 1/CLUL1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 5% cross-reactivity with recombinant human Clusterin is observed.
Human
IgG
Western Blot
LYOPH
Western Blot 0.1 ug/mL
CLUL1, Clusterin like 1, clusterin-like 1 (retinal), clusterin-like protein 1, RA337M, Retinal-specific clusterin-like protein
Goat
Affinity Purified
RUO
27098
Reconstitute at 0.2 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.