missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human CLIP3 (aa 345-432) Control Fragment Recombinant Protein
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Description
Highest antigen sequence indentity to the following orthologs: Mouse (97%), Rat (97%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-140025 (PA5-140025. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
Functions as a cytoplasmic linker protein. Involved in TGN-endosome dynamics.
Specifications
Specifications
| Accession Number | Q96DZ5 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 25999 |
| Name | Human CLIP3 (aa 345-432) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | 1500005P14Rik; AI844915; CAP-GLY domain containing linker protein 3; CAP-Gly domain-containing linker protein 3; clip-170 related protein 59 kDa; CLIP-170-related 59 kDa protein; Clip3; CLIPR59; Clipr-59; Cytoplasmic linker protein 170-related 59 kDa protein; restin-like 1; RGD1306245; RSNL1 |
| Common Name | CLIP3 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction