missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human CLIF (aa 465-526) Control Fragment Recombinant Protein

Product Code. 30209958
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30209958

Brand: Invitrogen™ RP105354

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (75%), Rat (75%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-84690 (PA5-84690. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

This gene encodes a basic helix-loop-helix transcription factor belonging to the PAS (PER, ARNT, SIM) superfamily. The PAS proteins play important roles in adaptation to low atmospheric and cellular oxygen levels, exposure to certain environmental pollutants, and diurnal oscillations in light and temperature. This protein forms a transcriptionally active heterodimer with the circadian CLOCK protein, the structurally related MOP4, and hypoxia-inducible factors, such as HIF1alpha. Consistent with its role as a biologically relevant partner of circadian and hypoxia factors, this protein is coexpressed in regions of the brain such as the thalamus, hypothalamus, and amygdala. Alternatively spliced transcript variants encoding different isoforms have been described for this gene. [provided by RefSeq, Oct 2011].
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q8WYA1
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 56938
Name Human CLIF (aa 465-526) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 4632430A05Rik; Arnt4; Arntl2; aryl hydrocarbon receptor nuclear translocator like 2; aryl hydrocarbon receptor nuclear translocator-like 2; aryl hydrocarbon receptor nuclear translocator-like protein 2; Basic-helix-loop-helix-PAS protein MOP9; BHLHE6; BMAL2; brain and muscle ARNT-like 2; brain and muscle Arnt-like protein 2 variant d; brain-muscle-ARNT-like protein 2; class E basic helix-loop-helix protein 6; CLIF; CYCLE-like factor; member of PAS protein 9; MGC149671; MGC149672; MOP9; PAS domain-containing protein 9; PASD9; transcription factor BMAL2
Common Name CLIF
Gene Symbol ARNTL2
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence IVSVNTLVLGHSEPGEASFLPCSSQSSEESSRQSCMSVPGMSTGTVLGAGSIGTDIANEILD
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.