missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human CLEC4D (aa 57-166) Control Fragment Recombinant Protein

Product Code. 30197598
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30197598

Brand: Invitrogen™ RP88578

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (65%), Rat (65%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-52131 (PA5-52131. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

This gene encodes a member of the C-type lectin/C-type lectin-like domain (CTL/CTLD) superfamily. Members of this family share a common protein fold and have diverse functions, such as cell adhesion, cell-cell signalling, glycoprotein turnover, and roles in inflammation and immune response. This gene is closely linked to other CTL/CTLD superfamily members on chromosome 12p13 in the natural killer gene complex region.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q8WXI8
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 338339
Name Human CLEC4D (aa 57-166) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias CD368; Clec4d; CLEC6; CLEC-6; CLECSF8; CLECSF8Mpcl; C-type (calcium dependent, carbohydrate recognition domain) lectin, superfamily member 8; C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamily member 8; C-type lectin domain family 4 member D; C-type lectin domain family 4, member d; C-type lectin receptor; C-type lectin superfamily member 8; C-type lectin, superfamily member 8; C-type lectin-like receptor 6; macrophage C-type lectin; macrophage-restricted C-type lectin; Mcl; MGC40078; Mpcl; superfamilymember 8
Common Name CLEC4D
Gene Symbol CLEC4D
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence KLEHHAKLKCIKEKSELKSAEGSTWNCCPIDWRAFQSNCYFPLTDNKTWAESERNCSGMGAHLMTISTEAEQNFIIQFLDRRLSYFLGLRDENAKGQWRWVDQTPFNPRR
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.