missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human CLEC2D (aa 60-160) Control Fragment Recombinant Protein

Product Code. 30211551
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30211551

Brand: Invitrogen™ RP89969

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (47%), Rat (47%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-53617 (PA5-53617. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Recently, Zhou et al have identified a protein that can inhibit multinucleate Osteoclast formation in murine osteoblast and spleen cell co-cultures. This protein called, Osteoclast Inhibitory Lectin (OCIL) can also inhibit osteoclast formation in spleen cell cultures treated with RANKL and M-CSF. Furthermore, mOCIL acted directly on macrophage/monocyte cells as evidenced by its inhibitory action on adherent spleen cell cultures, which were depleted of stromal and lymphocytic cells. mOCIL completely inhibited osteoclast formation during the proliferative phase of osteoclast formation and resulted in 70% inhibition during the differentiation phase. Osteoblast OCIL mRNA expression was enhanced by parathyroid hormone, calcitriol, interleukin-1 and IL-11, and retinoic acid. OCIL is expressed in osteoblasts and chondrocytes as well as in a variety of extraskeletal tissues. Since OCIL expression was found to be very similar to distribution for RANKL, it has been suggested that RANKl and OCIL may interact in skeleton and in extracellular tissues.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9UHP7
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 29121
Name Human CLEC2D (aa 60-160) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias CLAX; Clec2d; Clrb; Clr-b; C-type lectin domain family 2 member D; C-type lectin domain family 2, member d; C-type lectin related f; C-type lectin superfamily 2, member D; C-type lectin-domain family 2 member D; C-type lectin-related protein B; lectin-like NK cell receptor; Lectin-like transcript 1; lectin-like transmembrane protein; LLT1; LLT-1; MGC123478; OCIL; Osteoclast inhibitory lectin
Common Name CLEC2D
Gene Symbol CLEC2D
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence RANCHQEPSVCLQAACPESWIGFQRKCFYFSDDTKNWTSSQRFCDSQDADLAQVESFQELNFLLRYKGPSDHWIGLSREQGQPWKWINGTEWTRQFPILGA
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.