missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CLEC12B Alexa Fluor™ 405-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB11476V-100UG
This item is not returnable.
View return policy
Description
CLEC12B Monoclonal antibody specifically detects CLEC12B in Human samples. It is validated for Flow Cytometry
Specifications
| CLEC12B | |
| Monoclonal | |
| Alexa Fluor 405 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
| C-type lectin domain family 12 member B, C-type lectin domain family 12, member B, Macrophage Antigen H, UNQ5782, UNQ5782/PRO16089 | |
| Chinese Hamster Ovary cell line, CHO-derived human CLEC12B, Leu65-Asp276, Accession # Q2HXU8 | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry | |
| 1062714 | |
| Flow Cytometry | |
| Q2HXU8 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 387837 | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction