missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CLEC-1 Antibody, R&D Systems™

Goat Polyclonal Antibody
£167.00 - £501.00
Specifications
| Antigen | CLEC-1 |
|---|---|
| Concentration | LYOPH |
| Dilution | Western Blot 0.1 ug/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18778267
![]() |
R&D Systems
AF1704-SP |
25 μg |
£167.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18079201
![]() |
R&D Systems
AF1704 |
100 μg |
£501.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CLEC-1 Polyclonal specifically detects CLEC-1 in Human samples. It is validated for Western Blot.Specifications
| CLEC-1 | |
| Western Blot 0.1 ug/mL | |
| Polyclonal | |
| Goat | |
| Human | |
| CLEC1, CLEC-1, CLEC1A, CLEC1C-type lectin domain family 1 member A, C-type lectin domain family 1, member A, C-type lectin-like receptor 1, C-type lectin-like receptor-1, MGC34328 | |
| CLEC1A | |
| IgG | |
| Affinity Purified | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| LYOPH | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Q8NC01 | |
| 51267 | |
| Mouse myeloma cell line NS0-derived recombinant human CLEC-1 Gln77-Asp280 Accession # Q8NC01 | |
| Primary | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Detects human CLEC-1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 1% cross-reactivity with recombinant human CLEC-2 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title