missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CLEC-1 Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB1704
This item is not returnable.
View return policy
Description
CLEC-1 Monoclonal specifically detects CLEC-1 in Human samples. It is validated for Western Blot.
Specifications
| CLEC-1 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| Q8NC01 | |
| CLEC1A | |
| Mouse myeloma cell line NS0-derived recombinant human CLEC-1 Gln77-Asp280 Accession # Q8NC01 | |
| 500 μg | |
| Primary | |
| Detects human CLEC-1 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human CLEC-2 is observed. | |
| Human | |
| Purified |
| Western Blot | |
| 219003 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CLEC1, CLEC-1, CLEC1A, CLEC1C-type lectin domain family 1 member A, C-type lectin domain family 1, member A, C-type lectin-like receptor 1, C-type lectin-like receptor-1, MGC34328 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 51267 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction