missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human CLCA3 Full-length ORF (AAI46528.1, 1 a.a. - 262 a.a.) Recombinant Protein with GST-tag at N-terminal

Product Code. 16193942
Change view
Click to view available options
Quantity:
10 μg
25 μg
Unit Size:
10µg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16193942 10 μg 10µg
16103952 25 μg 25µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16193942 Supplier Abnova™ Supplier No. H00009629P01.10ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used for AP, Array, ELISA, WB-Re

This gene is a transcribed pseudogene belonging to the calcium sensitive chloride conductance protein family. To date, all members of this gene family map to the same site on chromosome 1p31-p22 and share high degrees of homology in size, sequence and predicted structure, but differ significantly in their tissue distributions. This gene contains several nonsense codons compared to other family members that render the transcript a candidate for nonsense-mediated mRNA decay (NMD). Therefore, this gene is unlikely to be protein-coding. [provided by RefSeq]

Sequence: MVFSLKVILFLSLLLSPVLKSSLVTLNNNGYDGIVIAINPSVPEDEKLIQNIKEMVTEASTYLFHATKQRAYFRNVSILIPMTYKSKSEYLIPKQESYDQADVIVADLYLKYGDDPYTLQYGQCGDKGQYIHFTPNFLLTNNLATYGPRGRVFVHEWAHLRWGVFDEYNVDQPFYISRRNTTEATRCSTRITVYMVLNECKGASCIARPFRRDSKTGLYEAKCTFIPKRSQTAKESIVFMQNLDSVTEFCTEKTHNKEAPNL

Specifications

Accession Number AAI46528.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 9629
Molecular Weight (g/mol) 55.77kDa
Name CLCA3 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 μg
Immunogen MVFSLKVILFLSLLLSPVLKSSLVTLNNNGYDGIVIAINPSVPEDEKLIQNIKEMVTEASTYLFHATKQRAYFRNVSILIPMTYKSKSEYLIPKQESYDQADVIVADLYLKYGDDPYTLQYGQCGDKGQYIHFTPNFLLTNNLATYGPRGRVFVHEWAHLRWGVFDEYNVDQPFYISRRNTTEATRCSTRITVYMVLNECKGASCIARPFRRDSKTGLYEAKCTFIPKRSQTAKESIVFMQNLDSVTEFCTEKTHNKEAPNL
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Common Name CLCA3
Gene Symbol CLCA3
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.