missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Claudin-4 APC-conjugated Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems FAB4219A
This item is not returnable.
View return policy
Description
Claudin-4 Monoclonal specifically detects Claudin-4 in Human samples. It is validated for Flow Cytometry.
Specifications
| Claudin-4 | |
| Monoclonal | |
| APC | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| claudin 4, Claudin4, CLDN4, Clostridium perfringens enterotoxin receptor, Clostridium perfringens enterotoxin receptor 1, CPE-R, CPE-RCPETR, CPE-receptor, CPETR1, hCPE-R, WBSCR8, WBSCR8CPERCPETR1claudin-4, Williams-Beuren syndrome chromosomal region 8 protein | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 1364 | |
| Human | |
| Purified |
| Flow Cytometry | |
| 382321 | |
| Flow Cytometry 10 uL/10^6 cells | |
| NP_001296 | |
| CLDN4 | |
| NS0 mouse myeloma cell line transfected with human Claudin-4 Met1-Val209 Accession # NP_001296 | |
| 100 Tests | |
| Primary | |
| Detects human Claudin-4. Stains human Claudin-4 transfectants but not irrelevant transfectants. Recognizes an epitope in the first extracellular loop (aa Met29-Arg81). | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction