missing translation for 'onlineSavingsMsg'
Learn More

Human Claudin-4 Antibody, R&D Systems™
GREENER_CHOICE

Mouse Monoclonal Antibody

£167.00 - £406.00

Specifications

Antigen Claudin-4
Clone 382321
Concentration LYOPH
Dilution Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready
Applications Flow Cytometry, CyTOF
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18744708
GREENER_CHOICE
View Documents
R&D Systems
MAB4219-SP
25 μg
£167.00
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18182231
View Documents
R&D Systems
MAB4219
100 μg
£406.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Claudin-4 Monoclonal specifically detects Claudin-4 in Human samples. It is validated for Flow Cytometry, CyTOF-ready.
Specifications

Specifications

Claudin-4
LYOPH
Flow Cytometry, CyTOF
Unconjugated
Mouse
Human
NP_001296
1364
NS0 mouse myeloma cell line transfected with human Claudin-4 Met1-Val209 Accession # NP_001296
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Detects human Claudin-4. Stains human Claudin-4 transfectants but not irrelevant transfectants. Recognizes an epitope in the first extracellular loop (aa Met29-Arg81).
382321
Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready
Monoclonal
Purified
RUO
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
claudin 4, Claudin4, CLDN4, Clostridium perfringens enterotoxin receptor, Clostridium perfringens enterotoxin receptor 1, CPE-R, CPE-RCPETR, CPE-receptor, CPETR1, hCPE-R, WBSCR8, WBSCR8CPERCPETR1claudin-4, Williams-Beuren syndrome chromosomal region 8 protein
CLDN4
IgG2a
Protein A or G purified from hybridoma culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Vidéos
FDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.